Learn More
Leica Biosystems LABSORB LARGE 12X12.5 600/CS
Supplier: Leica Biosystems 3803240
L'Absorbs are a strong highly absorbent, lint-free alternative to paper towels and gauze. The cloth-like softness and lint-free surface makes them ideal for use in cover slipping and slide preparation. L'Absorbs are perfect for cleaning up all liquids but are specially suited to alcohol and xylene which evaporate upon use, allowing L'Absorbs to be reused. L'Absorbs will readily soak up oils and liquid paraffin wax and can be used for various blotting techniques, from highly technical procedures to simple applications such as drying a block from ice during microtomy. This product is one of the most versatile and useful in the laboratory.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.