Learn More
NABERTHERM INC The muffle furnaces are for daily laboratory use. They stand out for their excellent workmanship and high level of reliability. Can come equipped with a flap door or lift door at no extra charge.
Supplier: NABERTHERM INC L404K1LN1480
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
The muffle furnaces are for daily lab use, outstanding workmanship, high level of reliability, non-classified fiber insulation, housing made of structured stainless steel, heating from two sides (three sides L24/11-LT60/12), either flap (L) or lift door (LT)