Learn More
Cole-Parmer Cole-Parmer Kynar Gas Sampling Bag, 18" x 18", On/Off valve
Supplier:  Cole-Parmer 0141014
Encompass_Preferred
All standard models have either an on/off valve (3⁄16'' nickel-plated brass hose barb) or a silicone septum in polypropylene housing; dual-valve models have both. Max temp: 160°F (71°C). Standard Kynar Bags Single-wide seam; available in 2-mil and 4-mil thick styles. Dual-Valve Kynar Bags Double-wide seamed bags have both on/off and septum valves for convenient mixing.
Catalog No. NC1511207