Learn More
Hach Company Storm Water Test Kit, Model SW-1
Supplier: Hach Company 2481300
The Storm Water Multi-Parameter Water Quality Test Kit is ideal for on-site or laboratory monitoring of storm sewer outflow and industrial discharge. The kit includes all necessary reagents and apparatus for monitoring detergents (0-1.2 mg/L), total chlorine (0-3.4 mg/L), copper (0-4 mg/L), and phenols (0-4 mg/L), among other parameters. Also included is a pH Pocket Pro Tester for quick pH measurements.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.