Learn More
Hach Company Intellical™ ISECL18101 Chloride (Cl⁻) Ion Selective Electrode (ISE) with Calibration Reagents Pack
Supplier: Hach Company ISECL181AP
The Intellical™ Chloride ISE Analysis Package contains all you need to start measuring chloride (Cl-) with your HQD meter instrument: The Intellical™ ISECL181 Chloride ISE with 1 m cable, 1 L bottles of 100 mg/L and 500mL of 1000 mg/L of Hach Chloride standard solutions, pack of 100 Hach chloride ionic strength adjustor (ISA) Powder Pillows.