Learn More
Hach Company HQ440D Laboratory Fluoride (F⁻) Ion Meter Package with ISEF121 Ion Selective Electrode
Supplier: Hach Company 8508000
Designed for water quality experts, the Hach HQ440D Laboratory Multi-Meter is an advanced laboratory meter that takes the guesswork out of measurements. HQD meters connect with smart Intellical electrodes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced HQD Laboratory Meter is designed to withstand years of continuous lab use. HQD meters feature a simple user interface that does not require manuals or training to operate. The Intellical ISEF121 is a digital, combination fluoride ion selective electrode (ISE) with a non-refillable, single junction reference and built-in temperature sensor. The ISEF121 electrode is intended for laboratory use. The ISEF121 is ideal for measuring fluoride concentrations in drinking water, wastewater, and general water quality applications. The ISEF121’s solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.