Learn More
Krackeler Scientific Gold Seal for Inlet Valve
Supplier: Krackeler Scientific 9251829652EA
Specifications
Seal, Inlet | |
For GC and GC/MS inlet liners | |
Inlet washer, O-rings | |
For use with total flow rates of greater than 200mL/min |