Learn More
Krackeler Scientific Sample Kit for Flame Ionization Detectors
![SDP](/content/dam/fishersci/glyphs/sdp.png)
For use with 7890/6890/6850 Flame Ionization Detectors.
Supplier: Krackeler Scientific 921871060170EA
Description
- For performance evaluation of FIDs
- Sample is used for the HP 5880, 5890 and 6890 with a FID or TCD
- Solution of 0.033% C14, C15, and C16 normal alkanes in hexane
- Quantity: 3 x 0.5 ampoules
Specifications
7890/6890/6850 Flame Ionization Detectors | |
Sample Kit |