Learn More
Newcomer Supply KP Marker Plus Marker Pen, 12/box for painted end of slides and cassettes
Supplier: Newcomer Supply 5724
Ultra fine-tip (diameter approximately 0.75mm), dark black ink pen. Write easily on microscope slides and plastic cassettes. Will go through the harshest histology chemistries and stay on. Excellent choice for the toughest applications.