Learn More
DWK Life Sciences Kimble™ Kontes™ NMR Polyethylene Sample Tube Pressure Caps for 5 and 10mm Tubes
SDP
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporization pressures
Supplier:  DWK Life Sciences 8970950061
 View more versions of this product
Catalog No. K897095-0061
Add to Cart
Description
Description
Specifications
Specifications
Pressure Cap
White
5 mm NMR Sample Tube
Polyethylene
Provides tight-fitting and stays in place even with a build-up of internal vaporization pressures