Learn More
DWK Life Sciences Kimble™ Kontes™ Brand Rotary Evaporator Glass Vial Connecting Adapters
Ideal for use with small samples
Supplier: DWK Life Sciences 7471302019
Description
Ideal for use with small samples
Connect standard rotary evaporator traps to vials with variety of GPI thread sizes. Typical receivers include Accuform™ 1 to 10mL vials (06-100C, 06-100E, 06-100F, 06-100G). Any standard vial with GPI threads can be used.
Specifications
Standard Taper Joint: 19/22 | |
Glass | |
20-400 |