Learn More
Cole-Parmer Koflo 3/8-40C-4-6-2 Clear PVC schedule 40 pipe mixer; 3/8" NPT(M), 6 elements
Supplier:  Cole-Parmer 0466985
Encompass_Preferred
Install these mixers directly into your process line to achieve a 100% mixed output—mixing elements are welded inside the pipe. All mixers have threaded ends. Choose PVC, 316 stainless steel, PVDF, or PFA. The clear PVC mixers are ideal for a visual check of your mixing processes. Models with removable elements are available on request—call our Application Specialists for details.
Catalog No. NC1349977