Learn More
Cole-Parmer Koflo 3/16-17 316 SS tube mixer; 3/16" OD; 17 elements
Supplier: Cole-Parmer 0466950
These mixers consist of a series of fixed right- and left-hand elements. Tube and mixing elements are made of 316 stainless steel—ideal for mixing high-viscosity fluids such as epoxies. Mixers have plain ends and accept standard tube fittings