Learn More
Electron Microscopy Sciences Kodak T-Max 400 Film, 135/36
Supplier:  Electron Microscopy Sciences 74170
Encompass_Preferred
Especially useful for dimly lighted subjects, fast action, extending the distance range, and flash pictures. Used in scientific and biomedical work, especially where fluorescence photography is required. Features high speed (El 400), very high sharpness, extremely fine grain, high resolving power. High degree of enlargement and shorter push-processing development time and total darkness. Recommended Kodak Developers: T-Max, D-76, HR-110, Microdol-X.
Catalog No. 50-268-22