Learn More
Electron Microscopy Sciences Kodak T-Max 400 Film, 135/24
Supplier: Electron Microscopy Sciences 74168
Especially useful for dimly lighted subjects, fast action, extending the distance range, and flash pictures. Used in scientific and biomedical work, especially where fluorescence photography is required. Features high speed (El 400), very high sharpness, extremely fine grain, high resolving power. High degree of enlargement and shorter push-processing development time and total darkness. Recommended Kodak Developers: T-Max, D-76, HR-110, Microdol-X.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.