Learn More
Electron Microscopy Sciences Kodak T-Max 100 Film, 135/36
Supplier: Electron Microscopy Sciences 74164
Especially useful for detailed subjects when maximum image quality is needed. Recommended for copying black and white photographs. Medium speed (EI-100) with extremely high sharpness, fine grain. Very high degree of enlargement, expanded exposure latitude. Recommended Kodak Developers: T-Max, D-76, HC-110, Microdol-X, Duraflo RT