Learn More
Electron Microscopy Sciences Kodak Stop Bath Indicator 16 OZ
Supplier: Electron Microscopy Sciences 74341
For general use with films, plates, or papers. Features built-in color indicator to signal exhaustion. Dilutes 1:64. Each package makes 8 gallons.