Learn More
Metrohm USA KN03 1 MOLAR 250ML
Supplier:  Metrohm USA 62310010
Encompass
Electrolyte solution KNO3 1 M (reference electrolyte for combined silver electrodes). 1mol/L 250 mL.
Catalog No. NC0255071