Learn More
Bioworld Kligler Iron Agar, 500 g
Supplier:  Bioworld 306200291
Encompass_Preferred
Kligler Iron AgarDifferential medium for the presumptive identification of the Enterobacteriaceae based upon the fermentaton of dextrose and lactose and hydrogen sulfide productionpH: 7.4Working Concentration:52 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: orange to red agarSoluble in boiling waterComposition (g/L):Pancreatic Digest of Casein: 10g Peptic Digest of Animal Tissue: 10gLactose: 10gDextrose;1gFerric Ammonium Citrate:0.5gSodium Chloride; 5gSodium Thiosulfate: 0.5gPhenol Red; 0.025gAgar : 15g
Catalog No. 50-254-306