Learn More
Electron Microscopy Sciences Kit Lab-Sorb 16 oz jar plus 25 large bags
Supplier: Electron Microscopy Sciences 6855209
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Lab-Sorb converts liquid waste to solid waste for easy disposal. LabSorb liquids to solids in seconds and the granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste. It reduces the possibility of accidentally spilling liquid waste in the lab and if spills do occur, use LabSorb for safe liquid containment during cleanup. Apply to any volume of liquid waste including 500 ml, 600 ml, 1L and 2L vessels. One jar of LabSorb can absorb up to 50 litres of liquid. Includes 16oz jar, 25 large bags.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.