Learn More
Thermo Scientific™ KingFisher Apex Purification Systems
The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source.
Supplier: Thermo Scientific™ A46943
Description
The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source. It moves magnetic beads, not liquids, through the main steps of sample preparation: bind, wash, and elute. This process provides excellent quality, reproducible results, and reduces hands-on time for sample processing.
- User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
- Provides versatile and high-speed purification with reproducible results
- Easy to use interface with touch screen empowering the user to control all details of the application and purification process
- BindIx PC Software for added convenience
- Extensive selection of validated protocols for different type of applications
- Cloud library for quick protocol retrieval
Specifications
Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate | |
8 | |
96 Samples/Run | |
>98% with 2.8 μm particle size | |
Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics | |
96 | |
96 | |
Max 23 hr. 59 min. | |
96 KF, 96 DW, 96 storage tube | |
56 cm | |
46 cm | |
56 kg | |
79 cm | |
1 year manufacturer warranty and 1 year of field service with PM |
KF 96 plate 15 to 200 μL, 96 DW plate 15 to 1000 μL, 96 storage tube 30 to 200 μL | |
96-well | |
KingFisher Apex with 96 Combi head, Extended Warranty Package | |
Down to +4°C | |
+4°C above ambient temperature up to +100°C | |
ca. >1 μm | |
96 in one frame, interchangeable | |
2 Pc. | |
22 in. | |
18 in. | |
123 lb. | |
31 in. | |
1 day at customer site |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.