Learn More
Kimberly Clark Kimtech™ Polaris™ Xtra Nitrile Exam Gloves
Catalog Number 17103194
This item is not returnable. View return policy
This item is not returnable. View return policy
Kimtech™ is your partner in driving progress. Tested against some of the harshest chemicals used in your work environment, Kimtech™ Polaris™ Exam Nitrile Gloves meet and exceed the industry standards for exam gloves.
  • As part of Kimberly Clark's 150-year-old legacy of innovation and partnership with the scientific community, we're proud to present our next step into the future— Kimtech™ Polaris™ Nitrile Gloves.
  • The proprietary high-quality nitrile formulation delivers the highest level of protection in the Kimtech™ exam glove portfolio and has been tested for use with chemotherapy drugs and fentanyl.
  • With an average tensile strength that is more than double the industry standard, Kimtech™ Polaris™ provides the strength and comfort needed for performance and productivity; it's even certified by US Ergonomics.
  • Eligible for recycling through The RightCycle™ Program by Kimberly-Clark Professional™, 100% of your non-hazardous Kimtech™ Polaris™ Nitrile Exam Gloves can be diverted from the landfill.
  • With protection, durability, and comfort you can count on, these gloves are built for science, made for you
  • Ambidextrous