Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ ValueWare™ Heavy-Wall Filtering Flasks
SCI_ED

Made with a heavier wall than a standard Erlenmeyer flask.

Supplier:  DWK Life Sciences 57060250/EMD

 View more versions of this product

Catalog No. S00115



Add to Cart

Description

Description

  • With side tubulation and capacity scale
  • Designed to take one atmosphere of vacuum or 29 in. (73.7cm) of mercury
  • Sizes 25 through 500mL have side hose connection designed to accept 0.31 in. (7.9mm) I.D. flexible tubing
  • Sizes 1000mL and larger accept 0.38 in. (9.66mm) I.D. tubing

Compliance

Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E1406, Type II, Class 1 specification requirements.

Specifications

Specifications

33.46 in.
250 mL
50 to 250 mL
6.1 in.
85 mm
0.78 cm
Borosilicate Glass
25 mL
15.5 cm
6
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit