Learn More
DWK Life Sciences Kimble™ Prelabeled Glass Blood Typing Mark-M-Tubes™
SDP
Made of borosilicate glass
Supplier:  DWK Life Sciences 60A10BZ1/DEL
 View more versions of this product
Catalog No. 22-037282
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
Explore available promotions
This item is not returnable. View return policy
Description
Description
Made of borosilicate glass
Fused labels make the written information permanent and easy to read. Tubes are prelabeled with the blood type, and a color-coded band with an area for patient identification. The marking area is placed to allow an unobstructed view of cell suspension.
Order Info
Available in two tube sizes: 10 x 75mm and 12 x 75mm.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass
Blood typing
Permanent Blood Type Label and Color-Coded Band