Learn More
DWK Life Sciences Kimble™ KIMAX™ Brand Filtering Flasks
Designed to afford maximum strength
Supplier: DWK Life Sciences 27050500
Description
- Heavy walled
- Necks tooled for uniform stopper fit with a thick flat flange
- Without side tabulation
- Durable, white ceramic enamel scale to indicate approximate volume
Certifications
ASTM Specification E1406, Type I requirements
Specifications
ASTM, E1406, Type I | |
500 mL | |
300 to 1000 mL | |
7.48 in. | |
10.4 cm |
4.09 in. | |
Borosilicate Glass | |
50 mL | |
19 cm | |
7 |