Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ Three Way without Tip O-Ring HI-VAC Valves

With PTFE plugs

Supplier:  DWK Life Sciences 8267000004

Catalog No. K826700-0004


Add to Cart

Alert:

Polyacetal knob is not recommended for use over 110°C.


Description

Description

  • Three way configuration
  • FKM o-rings
  • 0-4mm bore range
  • 9mm stem O.D.
  • 010 o-ring size

Specifications

Specifications

0 to 0.15 in.
0 to 0.4 cm
PTFE plug
010
3-way Valve
0.35 in.
0.9 cm
Borosilicate Glass
0 to 4 mm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit