Learn More
DWK Life Sciences Kimble™ Kontes™ Organic Chemistry Microscale LABSETS™ and Kits
With die cut foam insert
Supplier: DWK Life Sciences 7462100000
Specifications
USP Type I and ASTM E438 Type I, Class A | |
Threaded Starter 14/10 Kit | |
Claisen adapter, jacketed condenser, hickman distillation head, 2mL Craig recrystallization tube, Craig recrystallization plug, drying tube, 5mL pear shaped flask, 5mL micro vial, 20-400 compression cap, size 112 o-ring, e.P., septum, syringe |