Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ Microscale Organic Chemistry LABSETS™ and Kits Components

Components for microscale organic chemistry LABSETS

Supplier:  DWK Life Sciences 7480130000

 View more versions of this product

Catalog No. K748013-0000


Add to Cart

Description

Description

  • Microflex Kits feature special chaste connectors and specially tooled glassware components, providing great versatility
  • They fit easily into lab bench drawers

Specifications

Specifications

Reaction Tube
Microscale organic chemistry
Component part of Williamson standard organic chemistry labsets and kits
100/Pk.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit