Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Heavy Wall Round Bottom with Joint Boiling Flasks
Single neck flask
$72.50 - $76.00
Specifications
Material | Borosilicate Glass |
---|---|
Standard Taper Joint | 35/25 |
Bottom Shape | Round |
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
K601050-0236
|
DWK Life Sciences
6010500236 |
100 mL | 35/25 |
Case for $72.50
|
|
||||
K601050-0136
|
DWK Life Sciences
6010500136 |
50 mL | 35/25 |
Case for $76.00
|
|
||||
Description
KimCote™ plastic coating provides an added measure of safety in the event of breakage.
Specifications
Borosilicate Glass | |
35/25 | |
Round |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title