Learn More
Kimberly-Clark Professional™ Polypro Shoe Covers
Click to view available options
Standard pack: 12; Yellow
Description
- Made from one-layer spunbond fabric
- Traction strip on sole provides slip resistance
- Recommended in environments where there is no potential for fluid contact
Specifications
Size | X-Large |
Material | Polypropylene |
Coating | Polypropylene |
Color | Blue |
Includes | Traction |
Product Type | Shoe Covers |