Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ A80 Chemical Permeation and Jet Liquid Protection Apron
Click to view available options
Size: 36 x 36 in.
Description
- Made of Saranex 23-P film, laminated to a durable fabric, which provides a soft, cloth-like feel against the skin. One size fits most
- Undergone ASTM F1001 Penetration Testing and is ideal for use where workers may come into contact with hazardous chemical vapors such as chemical handling / mixing / cleanup, hazardous material handling and waste cleanup, petrochemicals, utilities and for industrial hazmat teams
- Features a neck loop and ties
- Use for work in utilities, hazardous waste cleanup, industrial hazmat teams, petrochemicals and chemical handling
Specifications
Type | Controlled Environments Apron |
Material | 23-P Film Laminate |
Color | White |
For Use With (Application) | Critical and Controlled Environments |
Dimensions (L x W) | 73.6 x 111.7 cm (29 x 44 in.) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction