Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ A20 Breathable Particle Protection Denim Blue Coveralls
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
- Reflex™ design with elastic back, wrists and ankles, and one-inch zipper flap
- Seamless front to provide more protection in the primary exposure areas
- Breathable, patented Microforce™ barrier SMS fabric
- Barrier protection against dry particulates
- Strong and abrasion-resistant
Compliance
NFPA 99 Compliant Antistatic
Specifications
Size | 4X-Large |
Material | SMS |
Color | Blue |
Ankle Style | Elastic |
Closure Type | Zipper |
Certifications/Compliance | NFPA 99 |
For Use With (Application) | Laboratories, Decontamination, Remediation, Pesticides, Heavy Dust Environments |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction