Learn More
Kimberly-Clark Professional™ KleenGuard™ Unispec II Safety Eyewear
Click to view available options
White
Description
- Metal-free design with screw-less hinges
- Clear glasses (with clear temples) allow maximum visible light transmission; ideal for indoor use
- Hard coated lenses for scratch resistance and longer life
- Polycarbonate lenses provide 99.9% UVA/UVB/UVC protection
- Meet the ANSI Z87.1+ standards for impact and are TAA compliant
Specifications
Certifications/Compliance | Meets ANSI Z87.1+ standards for high impact (16727-01 and 16727-02 meet ANSI Z87.1+2010 and 16727-03 meets ANSI Z87.1+2015) and certified to meet the requirements of CSA Z94.3 |
Lens Tint | Clear |
No. per Pack | 50/Pk. |