Learn More
Kimberly-Clark Professional™ Kimtech™ Sterile Sleeve
Click to view available options
Surgical Sleeve
Description
- Each universally sized sleeve is made with Evolution 4 SMS fabric
- Meets NFPA Class 1 and ISO 11810 for flame resistance and ASTM D4966 for abrasion resistance
- Double bagged
- Individually packaged
- Easy to don
Compliance
Meets NFPA Class 1 and ISO 11810 for flame resistance; ASTM™ D4966 for abrasion resistance.
Specifications
Product Type | Sterile Sleeve |
Size | Universal |
Sterility | Sterile |
Disposable | Single-use |
Certifications/Compliance | NFPA Class 1, ISO 11810, ASTM D4966 |
Width (English) | 23.5 in. |