Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ KIMAX™ Pink Class A ôColorwareö Glass Flasks

Ideal for ease of identification in the lab

$109.60 - $540.00

Specifications

Material Borosilicate Glass
Color Pink
Class A
View More Specs

Products 6
Catalog Number Mfr. No. Capacity (Metric) Tolerance Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Tolerance Height (Metric) Price Quantity & Availability  
02-555-119
View Documents Promotion Details
DWK Life Sciences
28014PNK25
25 mL ±0.03 mL 118 mm
Case of 6 for $307.00
Add to cart
 
02-555-120
View Documents Promotion Details
DWK Life Sciences
28014PNK50
50 mL ±0.05 mm 148 mm
Case of 6 for $321.00
Add to cart
 
02-555-121
View Documents Promotion Details
DWK Life Sciences
28014PNK100
100 mL ±0.08 mL 180 mm
Case of 6 for $359.00
Add to cart
 
02-555-122
View Documents Promotion Details
DWK Life Sciences
28014PNK250
250 mL ±0.12 mL 250 mm
Case of 6 for $439.00
Add to cart
 
02-555-123
View Documents Promotion Details
DWK Life Sciences
28014PNK500
500 mL ±0.20 mL 287 mm
Case of 6 for $540.00
Add to cart
 
02-555-124
View Documents Promotion Details
DWK Life Sciences
28014PNK1000
1000 mL ±0.30 mL 338 mm
Case for $109.60
Add to cart
 
Description

Description

  • Flask with single graduation ring blasted on neck, calibrated to contain
  • Standard Taper ground glass stopper supplied, along with durable, color, matte finish marking area for use with an ordinary pencil
  • Replacement stopper is 850100
  • Designed from ASTM Specification E288, Class A requirements
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications

Specifications

Borosilicate Glass
Pink
A
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit