Learn More
Kimble™ KIMAX™ Heavy-duty Erlenmeyer Flasks with Wide Mouth and Tooled Lip
SDPSCI_ED
Supplier:  Kimble™ 26650500/EMD
 View more versions of this product
Catalog No. S340881
Add to Cart
This item is not returnable. View return policy
Description
Description
Our thick-walled, heavy-duty flasks are perfect for use on shakers
The wide-mouth design and tooled lip of this flask work extremely well with stoppers (Kimble 26650).
Specifications
Specifications
Top style: Heavy duty tooled with capacity scale
500 mL
100 to 500 mL
6.77 in.
Wide
10
3.8 in.
Borosilicate Glass
50 mL
17.2 cm
97 mm
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Accessories