Learn More
Kimble™ KIMAX™ Heavy-duty Erlenmeyer Flasks with Wide Mouth and Tooled Lip
SDPSCI_ED
Supplier:  Kimble™ 26650125/EMD
 View more versions of this product
Catalog No. S340861
Add to Cart
Description
Description
Our thick-walled, heavy-duty flasks are perfect for use on shakers
The wide-mouth design and tooled lip of this flask work extremely well with stoppers (Kimble 26650).
Specifications
Specifications
Top style: Heavy duty tooled with capacity scale
125 mL
50 to 125 mL
4.25 in.
Wide
6
2.59 in.
Borosilicate Glass
25 mL
10.8 cm
66 mm
Product Suggestions
Product Suggestions