Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ KHS 101 hydrochloride
Selective inducer of neuronal differentiation in hippocampal neural progenitors
$265.64 - $1097.36
Specifications
Purity | >98% |
---|---|
Product Type | KHS 101 hydrochloride |
Molecular Weight (g/mol) | 375.92 |
Inhibitors | KHS 101 hydrochloride |
Description
Selective inducer of neuronal differentiation; induces neuronal differentiation in cultured hippocampal neural progenitor cells (NPCs) by interacting with TACC3 (EC50 ∽1 μM). Suppresses astrocyte formation. Also induces acceleration of neuronal differentiation in the hippocampal dentate gyrus in vivo.Specifications
>98% | |
KHS 101 hydrochloride | |
375.92 | |
KHS 101 hydrochloride |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title