Learn More
Enzo Life Sciences Ketotifen. fumarate (1g). CAS: 34580-14-8
Supplier: Enzo Life Sciences BMLNH1060001
Selective histamine H1 antagonist. Inhibits histamine release from monocytes and lymphocytes at 0.1-10 µM. Purity: ≥98% (HPLC). Solubility: Soluble in water (10 mg/ml). Long Term Storage: Ambient.