Learn More
Haztech Systems, Inc HW6004 Ketone test pieces (10 ct.)
Supplier: Haztech Systems, Inc HW6004
HW6004 Ketone test pieces (10 ct.)Ketones: acetone, methyl ethyl ketone (MEK), cyclohexanone, acetophenone andmethyl isobutyl ketone and other solvents: ethyl acetate, phenylacetate, benzonitrile, aromatic hydrocarbon, ether, gasoline, diesel, tetrahydrofuran, benzonitrile, methylene chloride or perchloroethylene will attack the Ketone Test piece (positive test). Ketones (the most common) and acetates are often found in mixed solvents.Toluene and methylene chloride are also often found in mixed solvents and will give a positive test.Ethers are usually found as pure products.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.