Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kemtec™ Structures and Bridges Kit
Students gain a comprehensive understanding of structural engineering concepts for supporting or resisting loads while building structures and bridges.
Supplier: Aqua Phoenix Scientific 20100
Alert:
Choking hazard (1) Small parts. Not for children under 3 yrs.
Includes:
- Six sets of Engino Structures and Bridges parts
- Six Engino 36-page hands-on activity books
- Six sets of Engino 26-piece building instructions
- Six rulers
- Six packs of straws (100 count)
- Six pairs of scissors
- Spring scale
- String roll
- Masking tape
- Dowel rod
- Student worksheets (24 pages)
- Instructor manual with lesson plans
Requires : Impact goggles
Description
- Study different types of bridges and STEM principles that support their designs
- Construct working models of a house, suspension bridge, cable-stayed bridge, arch bridge and two truss bridges
- Challenge 1: Build the largest free standing tower possible with Engino™ components
- Challenge 2: Design and build a straw bridge that spans 70cm and will support a minimum 2kg distributed static load using 100 plastic straws and limited masking tape
- Grades 7 to 10
- Class size: 24 students
Specifications
Structures and Bridges Kit | |
Students will develop comprehensive knowledge of structural engineering and how it's used to support or resist loads by building models | |
24 Students |
7 to 14 | |
6 sets of engine structures and bridges parts, 6 engine 36page hands-on activity books, 6 sets of engine 26piece building instructions, 6 rulers, 6 packs of straws, 6 pairs of scissors, spring scale, string roll, masking tape, dowel rod, student worksheets, teacher's manual with lesson plans |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction