Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kemtec™ Stoichiometry Module
Explore types of chemical reactions, mole ratios and stoichiometric equivalents which are central to all chemical calculations.
Supplier: Aqua Phoenix Scientific 4456
Includes:
- Instructor Manual
- Student Manuals
- SDS
- Lab Quiz Sheets
- Toothpicks
- Pipettes
- Pencil Lead
- Forceps
- Empty Bottles with Caps
- Cotton String
- Microplates (24 Well)
- Microplates (96 Well)
- Copper Nitrate
- Nickel Nitrate
- Lead Nitrate
- Cobalt Nitrate
- Nitric Acid
- Aluminum Nitrate
- Iron Nitrate
- Sodium Carbonate
- Sodium Sulfate
- Sodium Chloride
- EDTA
- Sodium Iodide
- Sodium Thiocyanate
- Sodium Chromate
- Sodium Dichromate
- Sodium Hydroxide
- Calcium Nitrate
- Sodium Oxalate
- Copper Sulfate
- Hydrochloric Acid
- Nitric Acid
- Sulfuric Acid
Requires : Goggles and 9V batteries with clips and leads
This item is not returnable. View return policy
Description
Kit includes all required chemicals and supplies for performing experiments on a micro scale.
- Students perform series of experiments including electrochemistry decomposition reaction, single replacement reactions and double replacement reactions
- Students are encouraged to predict possible products, as well as write and balance reaction equations
- Students discover quantitative nature of chemical equations and experimental basis of balancing numerical coefficients
- Includes lab quiz
- Grades 9-12
- Class size: 24 students
Contains lead.
Specifications
Stoichiometry Microchemistry Module | |
Chemical reaction types, reaction of calcium nitrate, sodium oxalate, reaction of lead nitrate, sodium iodide |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction