Learn More
Kemtec™ Reflection of Light Kit
Study how light rays are reflected to give images back to our eyes.
Supplier: Aqua Phoenix Scientific 15100
Alert:
Choking hazard (1) Small parts. Not for children under 3 yrs.
Includes: Instructor's manual, students' manuals, cardboard bases, protractors, metric rulers, plane mirrors, straight pins, pencils, paper
Requires : Impact goggles
Description
- Rectilinear propagation of light, formation of images by plane mirrors
- Reflected image, virtual image, angle of incidence, and angle of reflection
- Extension activity includes construction of a simple periscope
- Includes instructor's manual, reproducible student guides, assessment questions
- Appropriate for any level physics course (including AP™) as well as general science courses
- Grades 9 to 14
- Class size: 24 students
Specifications
National standards | |
8 to 12 | |
Instructor manual, students manuals, cardboard bases, protractors, metric rulers, plane mirrors, straight pins, pencils, paper |
Reflection of Light Kit | |
Study of the basic laws of physics | |
12 Students |