Learn More
Kemtec™ Reef Restaurant Kit
Study effects of certain environmental conditions on coral reef and associated food webs.
Supplier: Aqua Phoenix Scientific 9112
Alert:
Choking hazard (1) Small parts. Not for children under 3 yrs.
Includes:
- Instructor Manual
- Student Data Sheets
- Colored Tums
- Garbage Bags
- Plastic Cups
- Wire
- Sea Salt
- Radish Seeds
- Petri Dishes
- Parafilm
Requires : Clear plastic 2L bottles, liquid fertilizer and small light sources
This item is not returnable. View return policy
Description
- Teaches students about natural resources, environmental quality, natural and human-induced hazards
- Includes two activities, allow three class periods
- Grades 5-9
- Class size: 24 students