Learn More
Kemtec™ AP™ Chemistry Kits: AP Chemistry Series Kits (16 Kits)

Order all 16 Kemtec AP Chemistry Kits at once.
$3066.00 - $3142.00
Specifications
Certifications/Compliance | Exceed 2013 AP next generation national standards |
---|---|
Grade | 10 to 14 |
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
S07345
![]() |
Aqua Phoenix Scientific
16130A |
Basic AP Chemistry Series Kit | Statistics, precision and accuracy, no Glassware, Spectrophotometric analysis of Copper, Beer's Law, synthesis and gravimetric analysis of Cobalt Oxalate Hydrate, Acid-Base titrations, an inquiry investigation, separating substances by absorption chromatography, stoichiometry, reactions with Copper compounds, stoichiometry, mole ratio of an unknown, determining molar mass by colligative properties, determining molar mass using the ideal gas equation, reaction rate and order, Determining molar enthalpy using Hess's law, Le Chatelier's principle featuring Ag+ reaction, identifying weak acids by pKa, preparing Buffer solutions and determining their properties, determining the equilibrium constant (and Temperature's Effect) with Beer's law, micro scale electrochemical cells voltaic and electrolytic |
Each for $3,066.00
|
|
||||
S07346
![]() |
Aqua Phoenix Scientific
16130B |
Enhanced AP Chemistry Series Kit | Statistics, precision and accuracy, with Glassware, Spectrophotometric analysis of Copper, Beer's Law, synthesis and gravimetric analysis of Cobalt Oxalate Hydrate, Acid-Base titrations, an inquiry investigation, separating substances by absorption chromatography, Stoichiometry, reactions with Copper compounds, stoichiometry, mole ratio of an unknown, determining molar mass by colligative properties, determining molar mass using the ideal gas equation, reaction rate and order, determining molar enthalpy using Hess's law, Le Chatelier's principle featuring Ag+ reaction, identifying weak acids by pKa, preparing Buffer solutions and determining their properties, Determining the Equilibrium Constant (and Temperature's Effect) with Beer's law, micro scale electrochemical cells voltaic and electrolytic |
Each for $3,142.00
|
|
||||
Description
Both series kits include a balance. And all kits are designed to exceed 2013 AP Next Generation National Standards.
The Basic AP Chemistry Series Kit includes:
- Statistics: Precision & Accuracy (no glassware S07328)
- Spectrophotometric Analysis of Copper: Beer's Law (S07330)
- Synthesis and Gravimetric Analysis of Cobalt Oxalate Hydrate (S07331)
- Acid-Base Titrations: An Inquiry Investigation (S07332)
- Separating Substances by Absorption Chromatography (S07333)
- Stoichiometry: Reactions with Copper Compounds (S07334)
- Stoichiometry: Mole Ratio of an Unknown (S07335)
- Determining Molar Mass by Colligative Properties (S07336)
- Determining Molar Mass Using the Ideal Gas Equation (S07337)
- Reaction Rate and Order (S07338)
- Determining Molar Enthalpy Using Hess's Law (S07339)
- Le Chatelier's Principle Featuring Ag+ reaction! (S07340)
- Identifying Weak Acids by pKa (S07341)
- Preparing Buffer Solutions and Determining Their Properties (S07342)
- Determining the Equilibrium Constant (and Temperature's Effect) with Beer's Law (S07343)
- Micro-Scale Electrochemical Cells – Voltaic and Electrolytic (S07344)
- Statistics: Precision & Accuracy (with glassware S07329)
- Spectrophotometric Analysis of Copper: Beer's Law (S07330)
- Synthesis and Gravimetric Analysis of Cobalt Oxalate Hydrate (S07331)
- Acid-Base Titrations: An Inquiry Investigation (S07332)
- Separating Substances by Absorption Chromatography (S07333)
- Stoichiometry: Reactions with Copper Compounds (S07334)
- Stoichiometry: Mole Ratio of an Unknown (S07335)
- Determining Molar Mass by Colligative Properties (S07336)
- Determining Molar Mass Using the Ideal Gas Equation (S07337)
- Reaction Rate and Order (S07338)
- Determining Molar Enthalpy Using Hess's Law (S07339)
- Le Chatelier's Principle Featuring Ag+ reaction! (S07340)
- Identifying Weak Acids by pKa (S07341)
- Preparing Buffer Solutions and Determining Their Properties (S07342)
- Determining the Equilibrium Constant (and Temperature's Effect) with Beer's Law (S07343)
- Micro-Scale Electrochemical Cells – Voltaic and Electrolytic (S07344)
Kemtec AP kits are compatible with optional data collection and simulations using TI-Nspire handhelds and software. Free activities can be found at www.fisheredu.com/TI.
Specifications
Exceed 2013 AP next generation national standards | |
10 to 14 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.