Learn More
Scientific Gear LLC The MKV-710B is a compact volumetric KF titrator, which can measure in the range of 1 ppm to 100% H2O precisely in the sample of petroleum, chemical, pharmaceutical, and food industry.
Supplier: Scientific Gear LLC MKV710B
The MKV-710B is a compact volumetric Karl Fischer titrator, which can measure in the range of 1 ppm to 100%H2O precisely in the sample of petroleum, chemical, pharmaceutical, and food industry. The equipped 10mL burette discharge precision is ±0.015mL and Repeatability is ±0.005mL. Required solvent volume is 30 to 100mL. Measurement data can be saved for up to 100. The RS-232C and USB interface are equipped to connect an optional printer, electronic balance, and optional data acquisition software (SOFT-CAP).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.