Learn More
Perkin Elmer US LLC KBr Powder - 100 g
Supplier: Perkin Elmer US LLC 04977043
Potassium bromide (KBr) powder is included in the General Purpose FT-IR Sampling Kit (L1365312) and Liquid and Mull FT-IR Sampling Kit (L1365311).