Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER REAGENT, PYRIDINE-FREE SINGLE SOLUTION, 5 mg/ml | 2.5 L | GFS CHEMICALS, INC. Item #: 99602 | Bulk #: 1600
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: GFS Chemicals Inc 99602
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
WATERMARK KARL FISCHER REAGENT, PYRIDINE-FREE SINGLE SOLUTION, 5 mg/ml | For volumetric analysis. Suitable for general purpose or aldehydes and ketones.