Learn More
Kappler™ Automated Digital Pressure Test Kit
Click to view available options
Digital Pressure Test Kit
Description
Includes
Fittings, hoses, universal adapter and complete set-up and operating instructions.
Requires
Air supply with a 1/4 inch MNPT fitting.
Test is performed automatically, allowing the user to handle other duties while the test is in process
- Digital readout for more accurate testing
- Large LCD touchscreen allows easy test criteria selection and data input
- Test data can be copied to the provided USB stick via the built-in USB port for record keeping
Specifications
Product Type | Digital Pressure Test Kit |