Learn More
Chemglass Life Sciences Closure, 38mm, Polypropylene
Supplier: Chemglass Life Sciences CLS150310
Closure, 38mm, Polypropylene. Delong style closure made of polypropylene. Closure offers two positions, open for gas exchange or closed for humidified environment. Internal drip ring minimizes contamination. Autoclavable to 121°C.